Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001159523.1 | NP_001152995.1 | 4 | 705 | |
NM_009168.2 | NP_033194.1 | 4 | 702 | ||
GenBank mRNA | AB018423.1 | - | 4 | 701 | |
AK142491.1 | - | 4 | 718 | ||
AK164639.1 | - | 4 | 638 | ||
BC103798.1 | - | 4 | 598 | 30288676 |