Gene Transcripts
- Gene Symbol
- Sphk1
- Entrez Gene ID
- 20698
- Gene Name
- sphingosine kinase 1
- Gene Aliases
- 1110006G24Rik, Sk1, Spk1
- Chromosome Location
- Chr. 11: 116529817 - 116536675 on Build GRCm38
- UniGene ID
- Mm.20944
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001172472.1 |
NP_001165943.1
|
5, 6 |
824 |
|
NM_001172473.1 |
NP_001165944.1
|
5, 6 |
710 |
|
NM_001172475.1 |
NP_001165946.1
|
5, 6 |
749 |
|
NM_011451.3 |
NP_035581.1
|
3, 4 |
481 |
|
NM_025367.6 |
NP_079643.2
|
5, 6 |
707 |
|
GenBank mRNA |
AF068748.1 |
- |
5, 6 |
749 |
|
AF068749.1 |
- |
3, 4 |
481 |
|
AF415213.1 |
- |
3, 4 |
379 |
|
AK136289.1 |
- |
4, 5 |
495 |
|
AK155332.1 |
- |
5, 6 |
809 |
|
AK160900.1 |
- |
5, 6 |
700 |
|
BC037710.1 |
- |
5, 6 |
622 |
4221357 |
View Full Product Details