Gene Transcripts

Gene Symbol
Sptbn2
Entrez Gene ID
20743
Gene Name
spectrin beta, non-erythrocytic 2
Gene Aliases
Spnb3, mKIAA0302
Chromosome Location
Chr. 19: 4711223 - 4752360 on Build GRCm38
UniGene ID
Mm.329668
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021287.1 NP_067262.1 16 3329
GenBank mRNA AK147671.1 - 16 3345
AK172931.1 - 6 1638
BC079860.1 - 16 3329 6834745

View Full Product Details