Gene Transcripts

Gene Symbol
Thrsp
Entrez Gene ID
21835
Gene Name
thyroid hormone responsive
Gene Aliases
S14
Chromosome Location
Chr. 7: 97412957 - 97417510 on Build GRCm38
UniGene ID
Mm.28585
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009381.2 NP_033407.1 2 654
GenBank mRNA AK077300.1 - 2 447
BC009165.1 - 2 654 2811774
BC053378.1 - 1 105

View Full Product Details