Gene Transcripts

Gene Symbol
Zfp52
Entrez Gene ID
22710
Gene Name
zinc finger protein 52
Gene Aliases
KRAB11, Zfp-52, Zfp76, mszf23-2, mszf24, mszf34, mszf75, zfec29
Chromosome Location
Chr. 17: 21516295 - 21562601 on Build GRCm38
UniGene ID
Mm.296100
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144515.2 NP_653098.2 5 1434
GenBank mRNA AK032299.1 - 5 1435
BC021527.1 - 5 1346 5353641

View Full Product Details