Gene Transcripts

Gene Symbol
Nek3
Entrez Gene ID
23954
Gene Name
NIMA (never in mitosis gene a)-related expressed kinase 3
Gene Aliases
Chromosome Location
Chr. 8: 22128283 - 22167004 on Build GRCm38
UniGene ID
Mm.41413
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001162947.1 NP_001156419.1 15 1521
NM_011848.4 NP_035978.2 16 1573
GenBank mRNA AF093416.1 - 15 1526
AF099066.1 - 16 1542

View Full Product Details