Gene Transcripts
- Gene Symbol
- Mpdu1
- Entrez Gene ID
- 24070
- Gene Name
- mannose-P-dolichol utilization defect 1
- Gene Aliases
- LEC35, SL15, Supl15h
- Chromosome Location
- Chr. 11: 69656697 - 69662649 on Build GRCm38
- UniGene ID
- Mm.89579
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001301711.1 |
NP_001288640.1
|
4, 5 |
501 |
|
NM_011900.4 |
NP_036030.2
|
4, 5 |
466 |
|
GenBank mRNA |
AB024713.1 |
- |
4, 5 |
394 |
|
AK040432.1 |
- |
4, 5 |
405 |
|
AK080233.1 |
- |
4, 5 |
460 |
|
AK089426.1 |
- |
4, 5 |
408 |
|
AK172020.1 |
- |
4, 5 |
427 |
|
AK180694.1 |
- |
2, 3 |
164 |
|
BC026776.1 |
- |
4, 5 |
388 |
4159355 |
BC048871.1 |
- |
4, 5 |
394 |
5364408 |
CV561188.1 |
- |
4, 5 |
452 |
|
View Full Product Details