Gene Transcripts

Gene Symbol
Naglu
Entrez Gene ID
27419
Gene Name
alpha-N-acetylglucosaminidase (Sanfilippo disease IIIB)
Gene Aliases
Chromosome Location
Chr. 11: 101070094 - 101077672 on Build GRCm38
UniGene ID
Mm.95452
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013792.2 NP_038820.2 1, 2 415
GenBank mRNA AK143546.1 - 1, 2 416
AK164125.1 - 1, 2 394
AK170758.1 - 1, 2 398
BC055733.1 - 1, 2 389 6411530
U85247.1 - 1, 2 384

View Full Product Details