Gene Transcripts

Gene Symbol
Nt5c
Entrez Gene ID
50773
Gene Name
5',3'-nucleotidase, cytosolic
Gene Aliases
Dnt, Dnt1, Umph-2, Umph2
Chromosome Location
Chr. 11: 115490425 - 115491876 on Build GRCm38
UniGene ID
Mm.390379
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015807.1 NP_056622.1 4 462
GenBank mRNA AF078840.1 - 4 462
AK007418.1 - 4 501
BC024368.1 - 4 465 5065485

View Full Product Details