Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_024499.1 | NP_078775.1 | 8, 9 | 774 | |
GenBank mRNA | AK039966.1 | - | 7, 8 | 769 | |
AK051005.1 | - | 8, 9 | 773 | ||
AK079168.1 | - | 8, 9 | 784 | ||
AK165557.1 | - | 8, 9 | 798 | ||
AK201798.1 | - | 1, 2 | 67 | ||
BC003836.1 | - | 8, 9 | 774 | 3486148 |