Gene Transcripts

Gene Symbol
Nrgn
Entrez Gene ID
64011
Gene Name
neurogranin
Gene Aliases
0710001B06Rik, AI838505, NG, NG/RC3, Pss1, R75334, RC3
Chromosome Location
Chr. 9: 37544493 - 37552745 on Build GRCm38
UniGene ID
Mm.335065
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022029.2 NP_071312.1 4 1242
GenBank mRNA AK158630.1 - 4 1229

View Full Product Details