Gene Transcripts

Gene Symbol
Emc3
Entrez Gene ID
66087
Gene Name
ER membrane protein complex subunit 3
Gene Aliases
0610039A15Rik, AI225901, AW260416, Pob, Tmem111
Chromosome Location
Chr. 6: 113514887 - 113531638 on Build GRCm38
UniGene ID
Mm.275989
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175101.3 NP_780310.1 2 323
GenBank mRNA AK020058.1 - 2 294
AK075595.1 - 2 324
AK167819.1 - 2 172
AK206599.1 - 2 69
AK210449.1 - 2 32
AK213319.1 - 2 69
AK218418.1 - 2 69
BC004641.1 - 2 319 3157147

View Full Product Details