Gene Transcripts
- Gene Symbol
- LTA4H
- Entrez Gene ID
- 4048
- Gene Name
- leukotriene A4 hydrolase
- Gene Aliases
- Chromosome Location
- Chr. 12: 96000753 - 96043520 on Build GRCh38
- UniGene ID
- Hs.524648
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000895.2 |
NP_000886.1
|
1, 2 |
305 |
|
NR_132659.1 |
|
1, 2 |
305 |
|
XM_005268871.1 |
XP_005268928.1
|
1, 2 |
295 |
|
XM_011538348.1 |
XP_011536650.1
|
1, 2 |
295 |
|
XM_011538349.2 |
XP_011536651.1
|
1, 2 |
295 |
|
GenBank mRNA |
AB209740.1 |
- |
1, 2 |
172 |
|
AK293630.1 |
- |
1, 2 |
285 |
|
AK308184.1 |
- |
1, 2 |
234 |
|
AK310522.1 |
- |
1, 2 |
270 |
|
BC032528.1 |
- |
1, 2 |
222 |
5502553 |
CR457068.1 |
- |
1, 2 |
163 |
|
DB019628.1 |
- |
1, 2 |
305 |
|
J02959.1 |
- |
1, 2 |
193 |
|
J03459.1 |
- |
1, 2 |
231 |
|
View Full Product Details