Gene Transcripts

Gene Symbol
Ogfod2
Entrez Gene ID
66627
Gene Name
2-oxoglutarate and iron-dependent oxygenase domain containing 2
Gene Aliases
1300006G11Rik, 5730405M13Rik, AU041636, AW552395
Chromosome Location
Chr. 5: 124112058 - 124115480 on Build GRCm38
UniGene ID
Mm.7050
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025671.2 NP_079947.1 7 928
GenBank mRNA AK004916.1 - 4 1591
AK004943.1 - 7 929
AK017497.1 - 7 928
AK018534.1 - 7 929
AK168946.1 - 7 846
BC022762.1 - 6 776
BC069872.1 - 7 931 4219293

View Full Product Details