Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025878.1 | NP_080154.1 | 7 | 594 | |
GenBank mRNA | AB049954.1 | - | 7 | 618 | |
AK002554.1 | - | 4 | 339 | ||
AK010250.1 | - | 7 | 601 | ||
AK076033.1 | - | 4 | 339 | ||
AK076044.1 | - | 4 | 339 | ||
AK088540.1 | - | 7 | 590 | ||
AK166597.1 | - | 7 | 615 | ||
AK167067.1 | - | 7 | 595 | ||
AK168140.1 | - | 7 | 610 | ||
BC021752.1 | - | 7 | 594 | 5136720 |