Gene Transcripts

Gene Symbol
MAN2A1
Entrez Gene ID
4124
Gene Name
mannosidase alpha class 2A member 1
Gene Aliases
AMan II, GOLIM7, MANA2, MANII
Chromosome Location
Chr. 5: 109689366 - 109869625 on Build GRCh38
UniGene ID
Hs.432822
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002372.3 NP_002363.2 8 2267
XM_011543395.2 XP_011541697.1 8 1706
XM_017009472.1 XP_016864961.1 8 1188
GenBank mRNA AK314090.1 - 8 1608
BC142656.1 - 8 1569 40148064
BC142696.1 - 8 1569 40148740
D63998.1 - 8 1725
U31520.1 - 8 1218

View Full Product Details