Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006739.3 | NP_006730.2 | 3 | 308 | |
XM_006724242.3 | XP_006724305.1 | 3 | 363 | ||
GenBank mRNA | AK122853.1 | - | 3 | 285 | |
AK223323.1 | - | 3 | 298 | ||
AK291136.1 | - | 3 | 289 | ||
AK298624.1 | - | 3 | 289 | ||
AK301121.1 | - | 298 | |||
AK308343.1 | - | 3 | 258 | ||
AK312533.1 | - | 3 | 288 | ||
BC000142.1 | - | 3 | 260 | 2900229 | |
BC003656.2 | - | 2 | 234 | 3542201 | |
BM787690.1 | - | 3 | 311 | ||
CR456517.1 | - | 3 | 248 | ||
D83986.1 | - | 3 | 296 | ||
X74795.2 | - | 2 | 236 |