Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172399.3 | NP_765987.2 | 4 | 1863 | |
GenBank mRNA | AK029210.1 | - | 4 | 1864 | |
AK049802.1 | - | 4 | 1888 | ||
BC047154.1 | - | 4 | 1837 | 6489681 | |
CK618056.1 | - | 1 | 409 |