Gene Transcripts

Gene Symbol
Ubac2
Entrez Gene ID
68889
Gene Name
ubiquitin associated domain containing 2
Gene Aliases
1190008A14Rik, AW554959, Phgdhl1
Chromosome Location
Chr. 14: 121878556 - 122021035 on Build GRCm38
UniGene ID
Mm.441151
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026861.2 NP_081137.2 3 314
GenBank mRNA AK049109.1 - 3 263
AK158177.1 - 3 263
BC024467.1 - 3 300 4975777
BY080555.1 - 3 314

View Full Product Details