Gene Transcripts

Gene Symbol
Mrps9
Entrez Gene ID
69527
Gene Name
mitochondrial ribosomal protein S9
Gene Aliases
2310002A08Rik, AW322516, Mrp-S9, S9mt, mg637
Chromosome Location
Chr. 1: 42851233 - 42905683 on Build GRCm38
UniGene ID
Mm.252982
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023514.4 NP_076003.3 9 937
GenBank mRNA AJ250193.1 - 5 406
AK009082.1 - 9 887
AK049465.1 - 8 1926
AK144656.1 - 9 895
AK150985.1 - 9 894
BC022587.1 - 4 375
BC051936.1 - 8 630
BC069980.1 - 7 531
BC116437.2 - 9 887 40104493

View Full Product Details