Gene Transcripts

Gene Symbol
Rabep2
Entrez Gene ID
70314
Gene Name
rabaptin, RAB GTPase binding effector protein 2
Gene Aliases
2610011A08Rik, AI462746, Fra
Chromosome Location
Chr. 7: 126428416 - 126445907 on Build GRCm38
UniGene ID
Mm.35467
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030566.2 NP_085043.2 6 1284
GenBank mRNA AK029188.1 - 4 642
AK089938.1 - 6 988
AK144836.1 - 6 970
AK157832.1 - 6 945
AK218661.1 - 1 24
BC004088.1 - 5 1030 3593722
BC015287.1 - 6 1292 4240442

View Full Product Details