Gene Transcripts

Gene Symbol
Slc39a5
Entrez Gene ID
72002
Gene Name
solute carrier family 39 (metal ion transporter), member 5
Gene Aliases
1810013D05Rik, 2010205A06Rik, Zip5
Chromosome Location
Chr. 10: 128395931 - 128401276 on Build GRCm38
UniGene ID
Mm.22983
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136237.1 NP_001129709.1 3 344
NM_028051.3 NP_082327.2 2 430
NM_028092.3 NP_082368.1 2 306
GenBank mRNA AK007473.1 - 2 431
AK008448.1 - 2 307
BC028990.1 - 3 305 5041383
CA492656.1 - 3 384

View Full Product Details