Gene Transcripts

Gene Symbol
Glipr1
Entrez Gene ID
73690
Gene Name
GLI pathogenesis-related 1 (glioma)
Gene Aliases
2410114O14Rik, RTVP-1, RTVP1, mRTVP-1
Chromosome Location
Chr. 10: 111984792 - 112002619 on Build GRCm38
UniGene ID
Mm.173790
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028608.3 NP_082884.1 1 327
GenBank mRNA AK010768.1 - 1 259
AK137557.1 - 1 274
AK155009.1 - 1 255
AK170051.1 - 1 274
BB869923.1 - 1 329
BC025083.1 - 1 251 3711757
BC098231.1 - 1 301 3325823

View Full Product Details