Gene Transcripts

Gene Symbol
Dusp18
Entrez Gene ID
75219
Gene Name
dual specificity phosphatase 18
Gene Aliases
4930527G07Rik, AI430870, BB028064, DSP18, DUSP20, LMWDSP20
Chromosome Location
Chr. 11: 3895139 - 3901299 on Build GRCm38
UniGene ID
Mm.32588
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173745.5 NP_776106.1 2 429
GenBank mRNA AK015917.1 - 2 413
AK079664.1 - 2 425
AK081916.1 - 2 388
BC020036.1 - 2 423 3990751

View Full Product Details