Gene Transcripts

Gene Symbol
Atp11b
Entrez Gene ID
76295
Gene Name
ATPase, class VI, type 11B
Gene Aliases
1110019I14Rik, ATPIF, ATPIR, mKIAA0956
Chromosome Location
Chr. 3: 35754062 - 35856276 on Build GRCm38
UniGene ID
Mm.239416
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029570.3 NP_083846.2 21 2731
GenBank mRNA AK043943.1 - 10 1380
AK129248.1 - 21 2723
AK154156.1 - 21 2737
AK155372.1 - 21 2735
BC026986.1 - 6 800
BC076603.1 - 21 2601 30629594

View Full Product Details