Gene Transcripts

Gene Symbol
Prss23
Entrez Gene ID
76453
Gene Name
protease, serine 23
Gene Aliases
2310046G15Rik, 4930479H08Rik, AI790574, Spuve
Chromosome Location
Chr. 7: 89507785 - 89527254 on Build GRCm38
UniGene ID
Mm.250438
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029614.3 NP_083890.2 2 402
GenBank mRNA AK009847.1 - 2 361
AK033671.1 - 2 378
AK034439.1 - 2 368
AK078518.1 - 2 371
AK156348.1 - 2 369
BC018517.1 - 2 420 3597686

View Full Product Details