Gene Transcripts

Gene Symbol
Osbpl9
Entrez Gene ID
100273
Gene Name
oxysterol binding protein-like 9
Gene Aliases
2600011I06Rik, AU015843, Orp-9
Chromosome Location
Chr. 4: 109061145 - 109202272 on Build GRCm38
UniGene ID
Mm.366315
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134791.2 NP_001128263.1 11, 12 1036
NM_133885.4 NP_598646.4 14, 15 1269
NM_173350.3 NP_775485.2 12, 13 1075
GenBank mRNA AK035658.1 - 12, 13 1024
AK075981.1 - 1, 2 54
AK131984.1 - 14, 15 1240
AK172010.1 - 12, 13 1062
BC021507.1 - 10, 11 897 5347121
BC023759.1 - 14, 15 1224
BC026927.1 - 12, 13 958 5134780
BC089163.1 - 11, 12 977 5367603

View Full Product Details