Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_133952.2 | NP_598713.2 | 13 | 1876 | |
GenBank mRNA | AK028912.1 | - | 13 | 1862 | |
AK047292.1 | - | 13 | 1823 | ||
AK047476.1 | - | 13 | 1823 | ||
AK166809.1 | - | 13 | 1859 | ||
AK200002.1 | - | 4 | 312 | ||
BC004717.1 | - | 13 | 1865 | 3582116 |