Gene Transcripts

Gene Symbol
Psme4
Entrez Gene ID
103554
Gene Name
proteasome (prosome, macropain) activator subunit 4
Gene Aliases
AA409398, AU041366, TEMO, mKIAA0077
Chromosome Location
Chr. 11: 30771328 - 30881150 on Build GRCm38
UniGene ID
Mm.240066
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134013.3 NP_598774.2 16 2165
GenBank mRNA CV557020.1 - 5 405

View Full Product Details