Gene Transcripts

Gene Symbol
HGF
Entrez Gene ID
3082
Gene Name
hepatocyte growth factor
Gene Aliases
DFNB39, F-TCF, HGFB, HPTA, SF
Chromosome Location
Chr. 7: 81699006 - 81770438 on Build GRCh38
UniGene ID
Hs.396530
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000601.5 NP_000592.3 4 639
NM_001010931.2 NP_001010931.1 4 639
NM_001010932.2 NP_001010932.1 4 639
NM_001010933.2 NP_001010933.1 4 639
NM_001010934.2 NP_001010934.1 4 639
XM_006715956.2 XP_006716019.1 5 602
XM_011516115.2 XP_011514417.1 5 602
XM_017012097.1 XP_016867586.1 5 602
XM_017012098.1 XP_016867587.1 5 602
GenBank mRNA AB208900.1 - 4 546
AK291674.1 - 4 574
AK313694.1 - 4 489
BC022308.1 - 4 488
BC063485.1 - 4 497
BC105797.1 - 4 579
BC130284.1 - 4 487 40146315
BC130286.1 - 4 487 40146317
BU533235.1 - 582
FJ830861.1 - 4 412
FJ830862.1 - 4 412
L02931.1 - 4 472
M29145.1 - 4 513
M55379.1 - 4 412
M60718.1 - 4 484
M73239.1 - 4 477
M73240.1 - 4 477
M77227.1 - 4 494
U46010.1 - 4 466
X16323.1 - 4 546
X57574.1 - 4 489

View Full Product Details