Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001282356.1 | NP_001269285.1 | 12 | 1518 | |
NM_001282357.1 | NP_001269286.1 | 11 | 1355 | ||
NM_052832.3 | NP_439897.1 | 11 | 1479 | ||
NM_134266.1 | NP_599028.1 | 11 | 1479 | ||
GenBank mRNA | AF331521.1 | - | 11 | 1449 | |
AJ413228.1 | - | 11 | 1448 | ||
AJ413229.2 | - | 11 | 1480 | ||
AJ413230.1 | - | 11 | 1448 | ||
AK122933.1 | - | 4 | 370 | ||
BC113866.1 | - | 10 | 1331 | 40034306 | |
BC114474.1 | - | 10 | 1319 | 40034303 |