Gene Transcripts

Gene Symbol
CBL
Entrez Gene ID
867
Gene Name
Cbl proto-oncogene
Gene Aliases
C-CBL, CBL2, FRA11B, NSLL, RNF55
Chromosome Location
Chr. 11: 119206276 - 119308149 on Build GRCh38
UniGene ID
Hs.504096
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005188.3 NP_005179.2 5 985
XM_011543057.2 XP_011541359.1 5 985
XM_017018511.1 XP_016874000.1 4 420
GenBank mRNA BC132733.1 - 5 914
BC136463.1 - 5 914 9020452
X57110.1 - 5 991

View Full Product Details