Gene Transcripts
- Gene Symbol
- ASUN
- Entrez Gene ID
- 55726
- Gene Name
- asunder, spermatogenesis regulator
- Gene Aliases
- C12orf11, GCT1, Mat89Bb, NET48, SPATA30
- Chromosome Location
- Chr. 12: 26905171 - 26938326 on Build GRCh38
- UniGene ID
- Hs.505077
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK001499.1 |
- |
2 |
288 |
|
View Full Product Details