Gene Transcripts

Gene Symbol
C11orf63
Entrez Gene ID
79864
Gene Name
chromosome 11 open reading frame 63
Gene Aliases
Chromosome Location
Chr. 11: 122881640 - 122963862 on Build GRCh38
UniGene ID
Hs.164705
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024806.3 NP_079082.2 3 1104
NM_199124.2 NP_954575.1 3 1104
XM_005271680.4 XP_005271737.1 3 965
XM_006718915.2 XP_006718978.1 3 839
XM_011543009.2 XP_011541311.1 3 1173
XM_017018338.1 XP_016873827.1 3 839
XM_017018339.1 XP_016873828.1 3 872
XM_017018340.1 XP_016873829.1 3 683
XM_017018341.1 XP_016873830.1 3 1454
XM_017018342.1 XP_016873831.1 3 839
XM_017018343.1 XP_016873832.1 3 839
GenBank mRNA AK027207.1 - 2 458
AK097534.1 - 3 949
AK291625.1 - 3 707
BC009820.2 - 2 510
BC068507.1 - 3 867 5298472

View Full Product Details