Gene Transcripts

Gene Symbol
DCST2
Entrez Gene ID
127579
Gene Name
DC-STAMP domain containing 2
Gene Aliases
Chromosome Location
Chr. 1: 155018513 - 155033781 on Build GRCh38
UniGene ID
Hs.591491
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144622.2 NP_653223.2 3 548
XM_011509188.2 XP_011507490.1 1 15
GenBank mRNA AK057496.1 - 3 548
BC111794.1 - 3 527

View Full Product Details