Gene Transcripts

Gene Symbol
SLC36A1
Entrez Gene ID
206358
Gene Name
solute carrier family 36 member 1
Gene Aliases
Dct1, LYAAT1, PAT1, TRAMD3
Chromosome Location
Chr. 5: 151411344 - 151556318 on Build GRCh38
UniGene ID
Hs.269004
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308150.1 NP_001295079.1 9 1177
NM_078483.3 NP_510968.2 9 1177
XM_005268386.1 XP_005268443.1 9 1147
XM_006714759.3 XP_006714822.1 9 1226
XM_011537580.2 XP_011535882.1 9 1867
XM_011537581.1 XP_011535883.1 10 1295
XM_011537583.2 XP_011535885.1 10 5732
XM_011537584.2 XP_011535886.1 9 1386
XM_011537585.1 XP_011535887.1 10 1242
XM_011537586.2 XP_011535888.1 9 1990
XM_011537587.2 XP_011535889.1 9 1246
XM_011537588.2 XP_011535890.1 9 1090
XM_011537589.2 XP_011535891.1 9 1998
XM_011537590.1 XP_011535892.1 9 1005
XM_011537591.1 XP_011535893.1 9 980
XM_011537592.2 XP_011535894.1 9 984
XM_011537593.2 XP_011535895.1 10 1240
XM_011537594.1 XP_011535896.1 11 1354
XM_011537595.2 XP_011535897.2 9 1873
XM_017009216.1 XP_016864705.1 9 1073
XM_017009217.1 XP_016864706.1 9 1177
XM_017009218.1 XP_016864707.1 8 959
XM_017009219.1 XP_016864708.1 9 1227
GenBank mRNA AF516142.1 - 8 969
AK057340.1 - 9 1147
AK309323.1 - 9 1149
AY162213.1 - 9 1082
AY227111.1 - 10 1097
AY227112.1 - 3 465
BC136437.1 - 9 1048 9020426
BC136438.1 - 9 1048 9020427
BX537963.1 - 9 1174
GU296670.1 - 8 954

View Full Product Details