Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_078483.3 | NP_510968.2 | 11 | 2738 | |
XM_005268386.1 | XP_005268443.1 | 11 | 2708 | ||
XM_006714759.3 | XP_006714822.1 | 11 | 2787 | ||
XM_011537580.2 | XP_011535882.1 | 11 | 3428 | ||
XM_011537581.1 | XP_011535883.1 | 12 | 2856 | ||
XM_011537583.2 | XP_011535885.1 | 12 | 7293 | ||
XM_011537584.2 | XP_011535886.1 | 11 | 2947 | ||
XM_011537585.1 | XP_011535887.1 | 12 | 2803 | ||
XM_011537586.2 | XP_011535888.1 | 11 | 3551 | ||
XM_011537587.2 | XP_011535889.1 | 11 | 2807 | ||
XM_011537588.2 | XP_011535890.1 | 11 | 2651 | ||
XM_011537589.2 | XP_011535891.1 | 11 | 3559 | ||
XM_011537590.1 | XP_011535892.1 | 11 | 2566 | ||
XM_011537591.1 | XP_011535893.1 | 11 | 2541 | ||
XM_011537592.2 | XP_011535894.1 | 11 | 2545 | ||
XM_011537593.2 | XP_011535895.1 | 12 | 2801 | ||
XM_011537594.1 | XP_011535896.1 | 13 | 2915 | ||
XM_017009216.1 | XP_016864705.1 | 11 | 2634 | ||
GenBank mRNA | AF516142.1 | - | 10 | 2530 | |
AK025102.1 | - | 1 | 310 | ||
BC136437.1 | - | 11 | 2609 | 9020426 | |
BC136438.1 | - | 11 | 2609 | 9020427 | |
BX537963.1 | - | 11 | 2735 |