Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_174978.2 | NP_777638.2 | 18 | 2486 | |
XM_011536690.2 | XP_011534992.1 | 19 | 2616 | ||
XM_017021247.1 | XP_016876736.1 | 19 | 2614 | ||
XM_017021248.1 | XP_016876737.1 | 17 | 2449 | ||
XM_017021249.1 | XP_016876738.1 | 15 | 2181 | ||
XM_017021250.1 | XP_016876739.1 | 14 | 2531 | ||
XM_017021251.1 | XP_016876740.1 | 14 | 2132 | ||
GenBank mRNA | AK098187.1 | - | 18 | 2470 | |
AL832219.1 | - | 18 | 2536 |