Gene Transcripts

Gene Symbol
CDK15
Entrez Gene ID
65061
Gene Name
cyclin dependent kinase 15
Gene Aliases
ALS2CR7, PFTAIRE2, PFTK2
Chromosome Location
Chr. 2: 201790324 - 201895550 on Build GRCh38
UniGene ID
Hs.348711
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001261435.1 NP_001248364.1 10 1084
NM_001261436.1 NP_001248365.1 10 1084
NM_139158.2 NP_631897.1 10 994
XM_005246781.3 XP_005246838.1 10 1084
XM_005246782.4 XP_005246839.1 11 1331
XM_011511650.2 XP_011509952.1 10 1084
XM_011511652.1 XP_011509954.1 10 1084
XM_011511654.2 XP_011509956.1 10 1084
XM_011511655.2 XP_011509957.1 7 1091
XM_011511656.2 XP_011509958.1 7 698
XM_011511657.2 XP_011509959.1 5 595
GenBank mRNA AB053308.1 - 10 948
AK091710.1 - 1 137
AK131512.1 - 9 1021
AK292434.1 - 11 1201
AK310050.1 - 12 1317
AX772791.1 - 10 948
BC038807.1 - 10 1042 5200755
BX647394.1 - 10 1047

View Full Product Details