Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_032554.3 | NP_115943.1 | 1 | 556 | |
GenBank mRNA | AF345568.1 | - | 1 | 454 | |
AF385431.1 | - | 1 | 556 | ||
AF385432.1 | - | 1 | 556 | ||
AK122659.1 | - | 1 | 818 | ||
AK289557.1 | - | 1 | 740 | ||
AK313955.1 | - | 1 | 332 | ||
AY507143.1 | - | 1 | 313 | ||
BC066881.1 | - | 1 | 425 | 6971816 | |
BC066882.1 | - | 1 | 425 | 6971819 | |
BC066883.1 | - | 1 | 425 | 6971820 | |
BC067484.1 | - | 1 | 425 | 6971817 | |
BC095506.1 | - | 1 | 310 | ||
EU809458.1 | - | 1 | 313 |