Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014819.4 | NP_055634.3 | 4 | 610 | |
XM_017010099.1 | XP_016865588.1 | 5 | 686 | ||
GenBank mRNA | AB007898.1 | - | 4 | 507 | |
AK291759.1 | - | 4 | 544 | ||
BC030826.1 | - | 4 | 506 | 4811738 | |
BM469517.1 | - | 4 | 630 | ||
BQ228553.1 | - | 4 | 517 |