Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004388.2 | NP_004379.1 | 3, 4 | 589 | |
GenBank mRNA | AK075423.1 | - | 3, 4 | 853 | |
AK298703.1 | - | 3, 4 | 540 | ||
AK314640.1 | - | 3, 4 | 555 | ||
BC126333.1 | - | 3, 4 | 565 | 8992049 | |
BC126335.1 | - | 3, 4 | 565 | 8992051 | |
M95767.1 | - | 3, 4 | 524 |