Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005206.4 | NP_005197.3 | 3 | 1161 | |
NM_016823.3 | NP_058431.2 | 3 | 1331 | ||
GenBank mRNA | AK291060.1 | - | 3 | 1126 | |
BC001718.1 | - | 3 | 1283 | 3353237 | |
BC008506.1 | - | 3 | 1314 | 3452137 | |
BC009837.1 | - | 3 | 1127 | 4040536 | |
D10656.1 | - | 3 | 1288 |