Gene Transcripts

Gene Symbol
UBE4A
Entrez Gene ID
9354
Gene Name
ubiquitination factor E4A
Gene Aliases
E4, UBOX2, UFD2
Chromosome Location
Chr. 11: 118359581 - 118399211 on Build GRCh38
UniGene ID
Hs.602967
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204077.1 NP_001191006.1 3 412
NM_004788.3 NP_004779.2 3 412
GenBank mRNA AF087968.1 - 3 351
AK296369.1 - 3 359
AL713515.1 - 3 411
BC030973.1 - 3 361
BC052643.1 - 3 370
BC062695.1 - 3 392
BC111417.1 - 3 386 40038776
BC112367.1 - 3 389 40038767
BU167020.1 - 3 369
D50916.1 - 3 349
DB068702.1 - 3 412

View Full Product Details