Gene Transcripts

Gene Symbol
Rspo2
Entrez Gene ID
239405
Gene Name
R-spondin 2 homolog (Xenopus laevis)
Gene Aliases
2610028F08Rik, AA673245, D430027K22, ftls
Chromosome Location
Chr. 15: 43020788 - 43171595 on Build GRCm38
UniGene ID
Mm.193274
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172815.3 NP_766403.1 2 956
GenBank mRNA AK049891.1 - 2 707
AK052457.1 - 1 1080
AK087485.1 - 2 957
BC052844.1 - 2 307

View Full Product Details