Gene Transcripts

Gene Symbol
Adam29
Entrez Gene ID
244486
Gene Name
a disintegrin and metallopeptidase domain 29
Gene Aliases
Chromosome Location
Chr. 8: 55870912 - 55906964 on Build GRCm38
UniGene ID
Mm.67684
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175939.3 NP_787953.2 2 1635
GenBank mRNA AY190759.1 - 2 1619
BC119089.1 - 2 1616 8733838
BC119091.1 - 2 1616 8733840

View Full Product Details