Gene Transcripts
- Gene Symbol
- Gpi1
- Entrez Gene ID
- 14751
- Gene Name
- glucose phosphate isomerase 1
- Gene Aliases
- Amf, Gpi, Gpi-1, Gpi-1r, Gpi-1s, Gpi-1t, Gpi1-r, Gpi1-s, Gpi1-t, Gpi1s, MF, NK, NK/GPI, Nlk, Org, Pgi, Phi
- Chromosome Location
- Chr. 7: 34201327 - 34230336 on Build GRCm38
- UniGene ID
- Mm.589
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_008155.4 |
NP_032181.2
|
1, 2 |
253 |
|
GenBank mRNA |
AK031081.1 |
- |
1, 2 |
158 |
|
AK133827.1 |
- |
1, 2 |
175 |
|
AK134890.1 |
- |
1, 2 |
187 |
|
AK137805.1 |
- |
1, 2 |
181 |
|
AK147124.1 |
- |
1, 2 |
216 |
|
AK150341.1 |
- |
1, 2 |
184 |
|
AK159838.1 |
- |
1, 2 |
157 |
|
AK164566.1 |
- |
1, 2 |
152 |
|
AK169681.1 |
- |
1, 2 |
182 |
|
AK171956.1 |
- |
1 |
874 |
|
BC086640.1 |
- |
1, 2 |
200 |
6832250 |
BC088995.1 |
- |
1, 2 |
160 |
6403790 |
BC157972.1 |
- |
1, 2 |
166 |
9007294 |
BC172124.1 |
- |
1, 2 |
166 |
9054410 |
BY287889.1 |
- |
1, 2 |
223 |
|
BY350616.1 |
- |
1, 2 |
253 |
|
M14220.1 |
- |
1, 2 |
182 |
|
View Full Product Details