Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001171052.1 | NP_001164523.1 | 7 | 685 | |
NM_001171053.1 | NP_001164524.1 | 7 | 685 | ||
NM_001171054.1 | NP_001164525.1 | 7 | 685 | ||
NM_054082.2 | NP_473423.1 | 7 | 682 | ||
GenBank mRNA | AF288138.1 | - | 7 | 636 | |
AK129318.1 | - | 7 | 614 | ||
AK145942.1 | - | 7 | 630 | ||
AK146721.1 | - | 7 | 638 | ||
AK146905.1 | - | 7 | 638 | ||
AK147197.1 | - | 7 | 628 | ||
AK154856.1 | - | 7 | 625 | ||
AK158866.1 | - | 6 | 548 | ||
BC022124.1 | - | 7 | 649 | 5097283 | |
BC099960.1 | - | 7 | 602 | 30023887 | |
CA751287.1 | - | 1 | 24 |