Gene Transcripts

Gene Symbol
Exoc8
Entrez Gene ID
102058
Gene Name
exocyst complex component 8
Gene Aliases
AI414418, EXO84, Exo84p, R74783, SEC84
Chromosome Location
Chr. 8: 124890299 - 124897705 on Build GRCm38
UniGene ID
Mm.347360
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198103.2 NP_932771.1 1 2598
GenBank mRNA BC034060.1 - 1 1126
BC057052.1 - 1 2598 6822067

View Full Product Details