Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001282621.1 | NP_001269550.1 | 1 | 415 | |
NM_006667.4 | NP_006658.1 | 1 | 415 | ||
GenBank mRNA | AK293618.1 | - | 1 | 335 | |
AK313956.1 | - | 1 | 373 | ||
BC034238.1 | - | 1 | 349 | 3863377 | |
CR456993.1 | - | 1 | 296 | ||
CX762728.1 | - | 1 | 412 | ||
HY166171.1 | - | 1 | 415 | ||
Y12711.2 | - | 1 | 374 |